Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-34A10
Entrez Gene ID
181852
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_B0213.16
Chromosome Location
Chr.V: 3970702 - 3973001 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071703.4 NP_504104.1 2 - 3 471 99

View Full Product Details