Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-8
Entrez Gene ID
178808
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_K09D9.8
Chromosome Location
Chr.V: 4005519 - 4007565 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071716.2 NP_504117.1 3 - 4 794 126

View Full Product Details