Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-4
Entrez Gene ID
191841
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_K09D9.6
Chromosome Location
Chr.V: 4010470 - 4011966 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071718.1 NP_504119.1 4 - 5 819 101

View Full Product Details