Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35A3
Entrez Gene ID
187209
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_K09D9.2
Chromosome Location
Chr.V: 4022609 - 4024482 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071720.6 NP_504121.1 2 - 3 364 80

View Full Product Details