Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-39
Entrez Gene ID
191946
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_F38H12.1
Chromosome Location
Chr.V: 4104598 - 4106244 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071740.1 NP_504141.1 3 - 4 667 93

View Full Product Details