Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- lgc-1
- Entrez Gene ID
-
188099
- Gene Name
- Ligand-Gated ion Channel
- Gene Aliases
-
CELE_T05B4.1
- Chromosome Location
-
Chr.V: 4135028 - 4138825 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071753.7 |
NP_504154.5 |
8 - 9 |
804 |
113 |
- Gene Symbol
- nhr-57
- Entrez Gene ID
-
178813
- Gene Name
- Nuclear hormone receptor family member nhr-57
- Gene Aliases
-
CELE_T05B4.2
- Chromosome Location
-
Chr.V: 4131561 - 4138073 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071752.7 |
NP_504153.1 |
13 - 14 |
2646 |
113 |
View Full Product Details