Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- pmt-2
- Entrez Gene ID
-
178854
- Gene Name
- Phosphoethanolamine MethylTransferase
- Gene Aliases
-
CELE_F54D11.1
- Chromosome Location
-
Chr.V: 4628580 - 4635263 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071847.7 |
NP_504248.1 |
3 - 4 |
513 |
117 |
- Gene Symbol
- soc-1
- Entrez Gene ID
-
178855
- Gene Name
- Multisubstrate adapter protein soc-1
- Gene Aliases
-
CELE_F41F3.2
- Chromosome Location
-
Chr.V: 4628740 - 4646933 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_071849.6 |
NP_504250.2 |
3 - 4 |
353 |
117 |
View Full Product Details