Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-74
Entrez Gene ID
185628
Gene Name
Serpentine Receptor, class X
Gene Aliases
CELE_F41F3.6
Chromosome Location
Chr.V: 4658949 - 4662500 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071854.4 NP_504255.3 1 - 2 196 134
NR_136380.1 3 - 4 372 134

View Full Product Details