Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-73
Entrez Gene ID
185629
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F41F3.7
Chromosome Location
Chr.V: 4663616 - 4665751 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071855.3 NP_504256.2 6 - 7 745 145

View Full Product Details