Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nex-4
Entrez Gene ID
183294
Gene Name
protein-coding
Gene Aliases
CELE_C37H5.1
Chromosome Location
Chr.V: 4851438 - 4857293 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322553.3 NP_001309625.1 2 - 3 238 96
NM_001380638.1 NP_001368593.1 2 - 3 280 96
NM_001383302.1 2 - 3 272 96

View Full Product Details