Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-32
Entrez Gene ID
191815
Gene Name
Serpentine receptor class delta-32
Gene Aliases
CELE_T19H12.5
Chromosome Location
Chr.V: 4865648 - 4870392 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071902.5 NP_504303.1 2 - 3 385 124

View Full Product Details