Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-12
Entrez Gene ID
178880
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_T19H12.9
Chromosome Location
Chr.V: 4887165 - 4889629 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071908.7 NP_504309.1 2 - 3 410 101

View Full Product Details