Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-11
Entrez Gene ID
178882
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_T19H12.10
Chromosome Location
Chr.V: 4892588 - 4895071 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071910.4 NP_504311.1 5 - 6 1548 133

View Full Product Details