Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-23
Entrez Gene ID
191849
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C02E7.5
Chromosome Location
Chr.V: 4921102 - 4922616 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071921.2 NP_504322.1 2 - 3 428 152

View Full Product Details