Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-8
Entrez Gene ID
182575
Gene Name
Na/Ca eXchangers
Gene Aliases
CELE_C13D9.7
Chromosome Location
Chr.V: 4983747 - 4987197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071940.7 NP_504341.1 3 - 4 475 101

View Full Product Details