Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-7
Entrez Gene ID
182577
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_C13D9.9
Chromosome Location
Chr.V: 4997021 - 5000566 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071942.4 NP_504343.2 4 - 5 736 153

View Full Product Details