Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-46
Entrez Gene ID
178897
Gene Name
Uncharacterized protein
Gene Aliases
CELE_C04F5.3
Chromosome Location
Chr.V: 5097075 - 5099025 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071964.5 NP_504365.1 4 - 5 530 113

View Full Product Details