Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-38
Entrez Gene ID
182137
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C03A7.5
Chromosome Location
Chr.V: 5153906 - 5155616 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071979.3 NP_504380.2 4 - 5 737 109

View Full Product Details