Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-64
Entrez Gene ID
182140
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_C03A7.9
Chromosome Location
Chr.V: 5168248 - 5169686 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071985.6 NP_504386.2 2 - 3 283 75

View Full Product Details