Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-70
Entrez Gene ID
181870
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_B0238.6
Chromosome Location
Chr.V: 5258036 - 5259548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072005.1 NP_504406.1 4 - 5 820 144

View Full Product Details