Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-14A5
Entrez Gene ID
178926
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F08F3.7
Chromosome Location
Chr.V: 5421018 - 5423488 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072034.5 NP_504435.1 5 - 6 1340 152

View Full Product Details