Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glb-6
Entrez Gene ID
182780
Gene Name
Globin-like protein 6
Gene Aliases
CELE_C18C4.9
Chromosome Location
Chr.V: 5568184 - 5570533 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072068.6 NP_504469.1 3 - 4 335 68

View Full Product Details