Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pas-6
Entrez Gene ID
178943
Gene Name
Proteasome subunit alpha type-1
Gene Aliases
CELE_CD4.6
Chromosome Location
Chr.V: 5583161 - 5584343 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072071.6 NP_504472.1 4 - 5 558 75

View Full Product Details