Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hex-2
Entrez Gene ID
178951
Gene Name
Glyco_hydro_20 domain-containing protein
Gene Aliases
CELE_C14C11.3
Chromosome Location
Chr.V: 5654449 - 5658704 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072088.7 NP_504489.3 1 - 2 81 95
GenBank mRNA AM748821.2 - 1 - 2 52 95

View Full Product Details