Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-76
Entrez Gene ID
191870
Gene Name
protein-coding
Gene Aliases
CELE_F26F12.6
Chromosome Location
Chr.V: 5833831 - 5835183 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028689.4 NP_001023860.1 1 - 2 332 145

View Full Product Details