Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-18
Entrez Gene ID
182200
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C04E6.2
Chromosome Location
Chr.V: 5922536 - 5924297 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072142.2 NP_504543.1 1 - 2 137 149

View Full Product Details