Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
acy-2
Entrez Gene ID
191607
Gene Name
Adenylyl CYclase
Gene Aliases
CELE_C10F3.3
Chromosome Location
Chr.V: 5976024 - 5981478 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072152.6 NP_504553.1 11 - 12 2076 96

View Full Product Details