Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aps-1
Entrez Gene ID
178989
Gene Name
AP complex subunit sigma
Gene Aliases
CELE_F29G9.3
Chromosome Location
Chr.V: 6017926 - 6026331 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383313.1 1 - 2 125 67

View Full Product Details