Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-17
Entrez Gene ID
178990
Gene Name
DeHydrogenases, Short chain
Gene Aliases
CELE_F29G9.6
Chromosome Location
Chr.V: 6026329 - 6034814 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047644.2 NP_001041109.1 5 - 6 561 94
NR_136375.1 8 - 9 1164 94

View Full Product Details