Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jamp-1
Entrez Gene ID
178993
Gene Name
protein-coding
Gene Aliases
CELE_R01B10.5
Chromosome Location
Chr.V: 6047897 - 6051259 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373030.1 NP_001360559.1 3 - 4 544 134
NM_072167.8 NP_504568.1 3 - 4 544 134
GenBank mRNA AF304119.1 - 3 - 4 544 134

View Full Product Details