Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-10
Entrez Gene ID
191844
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_Y38A10A.3
Chromosome Location
Chr.V: 6065507 - 6067140 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072170.2 NP_504571.2 2 - 3 918 162

View Full Product Details