Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exp-2
Entrez Gene ID
179003
Gene Name
protein-coding
Gene Aliases
CELE_F12F3.1
Chromosome Location
Chr.V: 6131400 - 6143970 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028621.5 NP_001023792.1 4 - 5 435 115
NM_001380661.1 NP_001367760.1 4 - 5 462 115
NM_001380662.1 NP_001367759.1 4 - 5 514 115
NM_001383316.1 5 - 6 552 115

View Full Product Details