Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrck-1
Entrez Gene ID
179013
Gene Name
protein-coding
Gene Aliases
CELE_K08B12.5
Chromosome Location
Chr.V: 6257390 - 6267287 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313212.2 NP_001300141.1 8 - 9 4204 69
NM_001313213.3 NP_001300142.1 4 - 5 3268 69
NM_001313214.3 NP_001300143.1 4 - 5 3274 69
NM_001313215.3 NP_001300144.1 2 - 3 2077 69
NM_072198.7 NP_504599.2 8 - 9 4120 69

View Full Product Details