Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gad-1
Entrez Gene ID
179027
Gene Name
Gastrulation defective protein 1
Gene Aliases
CELE_T05H4.14
Chromosome Location
Chr.V: 6431320 - 6433496 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072234.5 NP_504635.1 3 - 4 1130 78

View Full Product Details