Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
moc-2
Entrez Gene ID
189078
Gene Name
MoCF_biosynth domain-containing protein
Gene Aliases
CELE_W01A11.6
Chromosome Location
Chr.V: 6502368 - 6504181 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383320.1 4 - 5 286 71

View Full Product Details