Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dre-1
Entrez Gene ID
179045
Gene Name
protein-coding
Gene Aliases
CELE_K04A8.6
Chromosome Location
Chr.V: 6533617 - 6546500 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380672.1 NP_001368566.1 7 - 8 2391 82
NM_001380673.1 NP_001367735.1 7 - 8 2400 82

View Full Product Details