Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spp-19
Entrez Gene ID
186974
Gene Name
Activin_recp domain-containing protein
Gene Aliases
CELE_K04A8.9
Chromosome Location
Chr.V: 6560173 - 6561692 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072265.5 NP_504666.3 3 - 4 532 67

View Full Product Details