Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-46
Entrez Gene ID
179058
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_K11G9.4
Chromosome Location
Chr.V: 6679101 - 6681573 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072293.6 NP_504694.1 4 - 5 647 72

View Full Product Details