Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-30
Entrez Gene ID
189125
Gene Name
Serpentine receptor class gamma-30
Gene Aliases
CELE_W02F12.7
Chromosome Location
Chr.V: 6716805 - 6718332 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072301.5 NP_504702.3 3 - 4 572 125

View Full Product Details