Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grl-9
Entrez Gene ID
179066
Gene Name
Ground-like domain-containing protein
Gene Aliases
CELE_ZC487.4
Chromosome Location
Chr.V: 6730216 - 6733084 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072305.5 NP_504706.1 3 - 4 992 60

View Full Product Details