Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spe-39
Entrez Gene ID
179071
Gene Name
protein-coding
Gene Aliases
CELE_ZC404.3
Chromosome Location
Chr.V: 6785451 - 6787797 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383328.1 1 - 2 136 78

View Full Product Details