Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-54
Entrez Gene ID
188525
Gene Name
Ligand-Gated ion Channel
Gene Aliases
CELE_T15B7.16
Chromosome Location
Chr.V: 6840217 - 6846607 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356670.2 NP_001343648.1 8 - 9 1233 106

View Full Product Details