Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-68
Entrez Gene ID
179080
Gene Name
protein-coding
Gene Aliases
CELE_K11C4.5
Chromosome Location
Chr.V: 6901681 - 6929038 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356673.4 NP_001343696.1 27 - 27 9891 79
NM_001356674.3 NP_001343811.1 27 - 27 9891 79
NM_001356675.3 NP_001343689.1 27 - 27 9891 79
NM_001356676.3 NP_001343809.1 27 - 27 9891 79
NM_001356677.4 NP_001343695.1 29 - 29 10239 79
NM_001356678.4 NP_001343810.1 29 - 29 10239 79
NM_001356679.3 NP_001343688.1 29 - 30 10328 79
NM_001356680.4 NP_001343808.1 29 - 29 10239 79
NM_001356681.3 NP_001343815.1 27 - 28 9885 79
NM_001356682.3 NP_001343819.1 27 - 28 9885 79
NM_001356683.3 NP_001343813.1 27 - 28 9885 79
NM_001356684.3 NP_001343817.1 27 - 28 9885 79
NM_001356685.3 NP_001343814.1 29 - 30 10233 79
NM_001356686.3 NP_001343818.1 29 - 30 10233 79
NM_001356687.3 NP_001343812.1 29 - 30 10233 79
NM_001356688.3 NP_001343816.1 29 - 30 10233 79

View Full Product Details