Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-68
Entrez Gene ID
179080
Gene Name
protein-coding
Gene Aliases
CELE_K11C4.5
Chromosome Location
Chr.V: 6901681 - 6929038 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356675.3 NP_001343689.1 30 - 30 10828 121
NM_001356676.3 NP_001343809.1 30 - 30 10828 121
NM_001356679.3 NP_001343688.1 33 - 34 11265 121
NM_001356680.4 NP_001343808.1 32 - 32 11176 121
NM_001356683.3 NP_001343813.1 31 - 32 10822 121
NM_001356684.3 NP_001343817.1 31 - 32 10822 121
NM_001356687.3 NP_001343812.1 33 - 34 11170 121
NM_001356688.3 NP_001343816.1 33 - 34 11170 121

View Full Product Details