Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-42
Entrez Gene ID
179084
Gene Name
Nuclear hormone receptor family member nhr-42
Gene Aliases
CELE_C33G8.6
Chromosome Location
Chr.V: 6998819 - 7000651 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072370.4 NP_504771.1 5 - 6 702 131

View Full Product Details