Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srab-4
Entrez Gene ID
183177
Gene Name
Serpentine Receptor, class AB (class A-like)
Gene Aliases
CELE_C33G8.5
Chromosome Location
Chr.V: 7004150 - 7005880 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072371.5 NP_504772.1 3 - 4 899 131

View Full Product Details