Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
odr-2
Entrez Gene ID
179098
Gene Name
protein-coding
Gene Aliases
CELE_T01C4.2
Chromosome Location
Chr.V: 7103543 - 7114439 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028918.4 NP_001024089.1 2 - 3 689 97
NM_001028919.5 NP_001024090.1 2 - 3 336 97
NM_001028920.4 NP_001024091.1 3 - 4 376 97
GenBank mRNA AF324050.1 - 3 - 4 365 97
AF324051.1 - 2 - 3 300 97
AF324052.1 - 2 - 3 457 97

View Full Product Details