Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-37
Entrez Gene ID
184293
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F10D2.6
Chromosome Location
Chr.V: 7141923 - 7145318 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072410.7 NP_504811.2 2 - 3 471 145

View Full Product Details