Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-39
Entrez Gene ID
3564933
Gene Name
protein-coding
Gene Aliases
CELE_F10D2.2
Chromosome Location
Chr.V: 7157142 - 7159235 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383331.1 1 - 2 254 129
NM_072415.4 NP_504816.2 1 - 2 307 129

View Full Product Details