Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-13
Entrez Gene ID
191935
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_R13D7.3
Chromosome Location
Chr.V: 7402655 - 7403910 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072495.1 NP_504896.1 4 - 5 568 131

View Full Product Details